siRNA Details
| Gene Symbol: Dhx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007842.2 | NP_031868.2 | 15 | 1861 | ||
| GenBank mRNA | AK135603.1 | - | 10 | 1176 | ||
| AK141824.1 | - | 15 | 1856 | |||
| AK148423.1 | - | 15 | 1857 | |||
| AY512925.1 | - | 11 | 1317 | |||
| BC089159.1 | - | 15 | 1850 | |||
| U91922.1 | - | 15 | 1818 | |||
| U92080.1 | - | 4 | 556 | |||

Dhx9
Genomic Map