siRNA Details
| Gene Symbol: Cyp3a41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105159.1 | NP_001098629.1 | 12 | 1439 | ||
| GenBank mRNA | BC138083.1 | - | 1368 | 8861102 | ||
| BC138084.1 | - | 1366 | 8861103 | |||
| Gene Symbol: Cyp3a41a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017396.3 | NP_059092.2 | 12 | 1435 | ||
| GenBank mRNA | AB033414.1 | - | 12 | 1433 | ||

Cyp3a41b
Genomic Map