siRNA Details
| Gene Symbol: Orai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178751.3 | NP_848866.2 | 3 | 1416 | ||
| GenBank mRNA | AK036013.1 | - | 3 | 1406 | ||
| AK042940.1 | - | 3 | 1417 | |||
| AK079619.1 | - | 3 | 1362 | |||
| AK087038.1 | - | 1706 | ||||
| BC066070.1 | - | 3 | 1363 | 6853893 | ||

Orai2
Genomic Map