siRNA Details
| Gene Symbol: Kazn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109684.1 | NP_001103154.1 | 2, 3 | 727 | ||
| NM_001109685.1 | NP_001103155.1 | 3, 4 | 745 | |||
| NM_144531.3 | NP_653114.3 | 2, 3 | 727 | |||
| GenBank mRNA | AK018497.1 | - | 3, 4 | 723 | ||
| AK052809.1 | - | 3, 4 | 746 | |||
| AK173090.1 | - | 2, 3 | 727 | |||
| BC022941.1 | - | 2, 3 | 731 | |||
| CD349067.1 | - | 2, 3 | 302 | |||

Kazn
Genomic Map