siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144868.3 | NP_659117.2 | 31 | 5889 | ||
| GenBank mRNA | AF237953.1 | - | 29 | 3906 | ||
| AK144114.1 | - | 7 | 1149 | |||
| AK220471.1 | - | 9 | 1787 | |||
| BC038056.1 | - | 26 | 3997 | |||
| BC054751.1 | - | 31 | 5168 | |||
| BC055289.1 | - | 32 | 5692 | |||
| BC096538.1 | - | 31 | 5889 | |||

Pcnxl3
Genomic Map