siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144868.3 | NP_659117.2 | 18 | 3808 | ||
| GenBank mRNA | AF237953.1 | - | 16 | 1825 | ||
| BC025838.1 | - | 12 | 1311 | |||
| BC038056.1 | - | 13 | 1916 | |||
| BC054751.1 | - | 18 | 3087 | |||
| BC055289.1 | - | 19 | 3611 | |||
| BC096538.1 | - | 18 | 3808 | |||

Pcnxl3
Genomic Map