siRNA Details
| Gene Symbol: Abcb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019552.2 | NP_062425.1 | 4 | 1208 | ||
| GenBank mRNA | AF266284.1 | - | 4 | 938 | ||
| AK081782.1 | - | 4 | 920 | |||
| BC046818.1 | - | 4 | 1164 | 5702696 | ||
| BC053020.1 | - | 4 | 1164 | 5702696 | ||
| BC054793.1 | - | 4 | 1208 | 6833785 | ||

Abcb10
Genomic Map