siRNA Details
| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034895.3 | NP_001030067.2 | 11 | 1714 | ||
| GenBank mRNA | AK033649.1 | - | 4 | 463 | ||
| AK122438.1 | - | 4 | 3322 | |||
| BC072571.1 | - | 10 | 2383 | |||
| BF449481.1 | - | 1 | 89 | |||

Zfr2
Genomic Map