siRNA Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081402.1 | NP_001074871.1 | 7 | 696 | ||
GenBank mRNA | AK159714.1 | - | 7 | 678 | ||
AK160933.1 | - | 7 | 631 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081402.1 | NP_001074871.1 | 7 | 696 | ||
GenBank mRNA | AK159714.1 | - | 7 | 678 | ||
AK160933.1 | - | 7 | 631 |