siRNA Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033315.2 | NP_001028487.2 | 1 | 683 | ||
| GenBank mRNA | AB354583.1 | - | 1 | 427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hipk4| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033315.2 | NP_001028487.2 | 1 | 683 | ||
| GenBank mRNA | AB354583.1 | - | 1 | 427 | ||