siRNA Details
| Gene Symbol: Smim13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135577.2 | NP_001129049.1 | 2 | 1619 | ||
| GenBank mRNA | BC024659.1 | - | 2 | 1291 | ||
| BC066054.1 | - | 2 | 1602 | |||
| BC094222.1 | - | 2 | 1601 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim13| Gene Symbol: Smim13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135577.2 | NP_001129049.1 | 2 | 1619 | ||
| GenBank mRNA | BC024659.1 | - | 2 | 1291 | ||
| BC066054.1 | - | 2 | 1602 | |||
| BC094222.1 | - | 2 | 1601 | |||