siRNA Details
| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081295.1 | NP_001074764.1 | 1 | 1246 | ||
| GenBank mRNA | AK087060.1 | - | 1 | 1023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef26| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081295.1 | NP_001074764.1 | 1 | 1246 | ||
| GenBank mRNA | AK087060.1 | - | 1 | 1023 | ||