siRNA Details
| Gene Symbol: Rab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293652.1 | NP_001280581.1 | 6 | 632 | ||
| NM_001293653.1 | NP_001280582.1 | 4 | 293 | |||
| NM_001293654.1 | NP_001280583.1 | 5 | 459 | |||
| NM_001293655.1 | NP_001280584.1 | 4 | 372 | |||
| NM_009005.3 | NP_033031.2 | 4 | 410 | |||
| GenBank mRNA | AB232599.1 | - | 3 | 204 | ||
| AK005004.1 | - | 4 | 372 | |||
| AK057671.1 | - | 4 | 339 | |||
| AK135659.1 | - | 5 | 460 | |||
| AK139914.1 | - | 4 | 363 | |||
| AK148117.1 | - | 4 | 403 | |||
| AK151078.1 | - | 4 | 384 | |||
| AK152666.1 | - | 6 | 607 | |||
| AK154234.1 | - | 4 | 391 | |||
| AK158971.1 | - | 4 | 294 | |||
| AK159507.1 | - | 4 | 340 | |||
| AK159928.1 | - | 4 | 311 | |||
| BC004597.1 | - | 4 | 328 | 3584891 | ||
| BC086793.1 | - | 4 | 326 | 6414012 | ||
| BY291181.1 | - | 4 | 411 | |||
| CT010352.1 | - | 3 | 204 | |||
| X89650.1 | - | 4 | 360 | |||

Rab7
Genomic Map