siRNA Details
| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 10 | 1659 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_003751234, XM_008769812, XM_006249680, XM_006249681, AB038495.

Cep170
Genomic Map