siRNA Details
| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 13 | 2905 | ||
| GenBank mRNA | BC038377.1 | - | 1 | 444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep170| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 13 | 2905 | ||
| GenBank mRNA | BC038377.1 | - | 1 | 444 | ||