siRNA Details
| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 13 | 2816 | ||
| GenBank mRNA | AK172953.1 | - | 1 | 295 | ||
| BC038377.1 | - | 1 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep170| Gene Symbol: Cep170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099637.2 | NP_001093107.2 | 13 | 2816 | ||
| GenBank mRNA | AK172953.1 | - | 1 | 295 | ||
| BC038377.1 | - | 1 | 355 | |||