siRNA Details
| Gene Symbol: Gal3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199366.4 | NP_955398.3 | 4 | 2293 | ||
| GenBank mRNA | AK144526.1 | - | 4 | 2294 | ||
| AK165692.1 | - | 4 | 2297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st2| Gene Symbol: Gal3st2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199366.4 | NP_955398.3 | 4 | 2293 | ||
| GenBank mRNA | AK144526.1 | - | 4 | 2294 | ||
| AK165692.1 | - | 4 | 2297 | |||