siRNA Details
| Gene Symbol: Ccdc171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081012.1 | NP_001074481.1 | 21 | 3570 | ||
| GenBank mRNA | AK037665.1 | - | 2 | 317 | ||
| AK039282.1 | - | 9 | 2266 | |||
| BC039810.1 | - | 3 | 409 | |||
| BC172165.1 | - | 20 | 3361 | |||

Ccdc171
Genomic Map