siRNA Details
| Gene Symbol: Cxx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018063.1 | NP_001018073.1 | 1 | 851 | ||
| GenBank mRNA | AK003646.1 | - | 1 | 830 | ||
| AK206042.1 | - | 1 | 219 | |||
| BC029838.1 | - | 1 | 851 | 3967024 | ||
| Gene Symbol: Cxx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024170.2 | NP_077132.1 | 1 | 883 | ||
| GenBank mRNA | AK002323.1 | - | 1 | 819 | ||
| BC003436.1 | - | 1 | 802 | 2647372 | ||

Cxx1b
Genomic Map