siRNA Details
| Gene Symbol: Avp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009732.1 | NP_033862.1 | 1 | 112 | ||
| GenBank mRNA | AK138179.1 | - | 1 | 145 | ||
| BC051997.1 | - | 1 | 125 | 5683720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avp| Gene Symbol: Avp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009732.1 | NP_033862.1 | 1 | 112 | ||
| GenBank mRNA | AK138179.1 | - | 1 | 145 | ||
| BC051997.1 | - | 1 | 125 | 5683720 | ||