siRNA Details
| Gene Symbol: Cnbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027585.2 | NP_081861.2 | 7 | 1032 | ||
| GenBank mRNA | AK014912.1 | - | 7 | 1055 | ||
| AK162372.1 | - | 7 | 913 | |||
| BC075727.1 | - | 4 | 616 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnbd2| Gene Symbol: Cnbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027585.2 | NP_081861.2 | 7 | 1032 | ||
| GenBank mRNA | AK014912.1 | - | 7 | 1055 | ||
| AK162372.1 | - | 7 | 913 | |||
| BC075727.1 | - | 4 | 616 | |||