siRNA Details
| Gene Symbol: Cnbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027585.2 | NP_081861.2 | 13 | 1799 | ||
| GenBank mRNA | AK014912.1 | - | 13 | 1822 | ||
| BC027204.1 | - | 13 | 1603 | 4037010 | ||
| BC140978.1 | - | 13 | 1782 | 9055590 | ||
| JN629039.1 | - | 13 | 1739 | |||

Cnbd2
Genomic Map