siRNA Details
| Gene Symbol: Gm5640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099302.1 | NP_001092772.1 | 4 | 1217 | ||
| Gene Symbol: Gm5936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081670.2 | NP_001075139.1 | 4 | 1213 | ||

Gm5640
Genomic Map