siRNA Details
Gene Symbol: Gm8909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081032.2 | NP_001074501.2 | 4 | 683 | ||
GenBank mRNA | AB267095.1 | - | 2 | 301 | ||
AB359231.2 | - | 4 | 683 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm8909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081032.2 | NP_001074501.2 | 4 | 683 | ||
GenBank mRNA | AB267095.1 | - | 2 | 301 | ||
AB359231.2 | - | 4 | 683 |