siRNA Details
| Gene Symbol: Gm14685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025383.2 | NP_001020554.1 | 4 | 2018 | ||
| GenBank mRNA | BC094343.1 | - | 4 | 1970 | 6402225 | |
| BC147366.1 | - | 1823 | 9056429 | |||
| BC151162.1 | - | 1823 | 9088051 | |||
| Gene Symbol: DXBay18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025384.3 | NP_001020555.2 | 5 | 2107 | ||
| GenBank mRNA | AK005439.1 | - | 5 | 2108 | ||

Gm14685
Genomic Map