siRNA Details
| Gene Symbol: Cd101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167906.1 | NP_001161378.1 | 9 | 3017 | ||
| GenBank mRNA | AM849328.1 | - | 9 | 3017 | ||
| AM849329.1 | - | 9 | 3017 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd101| Gene Symbol: Cd101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167906.1 | NP_001161378.1 | 9 | 3017 | ||
| GenBank mRNA | AM849328.1 | - | 9 | 3017 | ||
| AM849329.1 | - | 9 | 3017 | |||