siRNA Details
Gene Symbol: AY761184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 411 | ||
GenBank mRNA | AK008498.1 | - | 2 | 417 | ||
EU760895.1 | - | 1 | 207 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AY761184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 411 | ||
GenBank mRNA | AK008498.1 | - | 2 | 417 | ||
EU760895.1 | - | 1 | 207 |