siRNA Details
| Gene Symbol: Oxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130163.1 | NP_001123635.1 | 7 | 1198 | ||
| NM_001130165.1 | NP_001123637.1 | 7 | 1293 | |||
| NM_001130166.1 | NP_001123638.1 | 9 | 1557 | |||
| NM_130885.2 | NP_570955.1 | 7 | 1198 | |||
| GenBank mRNA | AF324899.1 | - | 7 | 1197 | ||
| AF333985.1 | - | 7 | 1198 | |||
| AK040881.1 | - | 7 | 1221 | |||
| AK052714.1 | - | 7 | 1363 | |||
| AK158703.1 | - | 9 | 1539 | |||

Oxr1
Genomic Map