siRNA Details
| Gene Symbol: Cep290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146009.2 | NP_666121.2 | 42 | 5982 | ||
| GenBank mRNA | AK029960.1 | - | 7 | 989 | ||
| AK172940.1 | - | 15 | 2638 | |||
| BC004690.1 | - | 8 | 1268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep290| Gene Symbol: Cep290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146009.2 | NP_666121.2 | 42 | 5982 | ||
| GenBank mRNA | AK029960.1 | - | 7 | 989 | ||
| AK172940.1 | - | 15 | 2638 | |||
| BC004690.1 | - | 8 | 1268 | |||