siRNA Details
| Gene Symbol: Adam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270996.1 | NP_001257925.1 | 9 | 1022 | ||
| NM_007404.2 | NP_031430.2 | 9 | 1022 | |||
| GenBank mRNA | AK155582.1 | - | 9 | 983 | ||
| AK161778.1 | - | 9 | 946 | |||
| BC052710.1 | - | 9 | 894 | |||

Adam9
Genomic Map