siRNA Details
| Gene Symbol: MESDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015154.2 | NP_055969.1 | 2, 3 | 541 | ||
| NR_126327.1 | 2, 3 | 541 | ||||
| GenBank mRNA | AK026606.1 | - | 2, 3 | 469 | ||
| AY359110.1 | - | 2, 3 | 527 | |||
| BC009210.2 | - | 2, 3 | 491 | 3637449 | ||
| BC012746.2 | - | 2, 3 | 491 | 3954629 | ||
| D42039.2 | - | 2, 3 | 457 | |||

MESDC2
Genomic Map