siRNA Details
| Gene Symbol: Serpina3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011458.2 | NP_035588.2 | 4 | 1026 | ||
| GenBank mRNA | AK004842.1 | - | 4 | 1023 | ||
| AK149436.1 | - | 4 | 1027 | |||
| X56786.1 | - | 4 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina3k| Gene Symbol: Serpina3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011458.2 | NP_035588.2 | 4 | 1026 | ||
| GenBank mRNA | AK004842.1 | - | 4 | 1023 | ||
| AK149436.1 | - | 4 | 1027 | |||
| X56786.1 | - | 4 | 1015 | |||