siRNA Details
| Gene Symbol: Gm10436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105254.1 | NP_001098724.1 | 4 | 1616 | ||
| GenBank mRNA | AK139576.1 | - | 3 | 1201 | ||
| BC094894.1 | - | 3 | 1178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10436| Gene Symbol: Gm10436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105254.1 | NP_001098724.1 | 4 | 1616 | ||
| GenBank mRNA | AK139576.1 | - | 3 | 1201 | ||
| BC094894.1 | - | 3 | 1178 | |||