siRNA Details
| Gene Symbol: Mcpt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010782.3 | NP_034912.3 | 2 | 1349 | ||
| GenBank mRNA | AF007120.1 | - | 1 | 13 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcpt9| Gene Symbol: Mcpt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010782.3 | NP_034912.3 | 2 | 1349 | ||
| GenBank mRNA | AF007120.1 | - | 1 | 13 | ||