siRNA Details
| Gene Symbol: G0s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008059.3 | NP_032085.1 | 2 | 732 | ||
| GenBank mRNA | AK003165.1 | - | 2 | 654 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G0s2| Gene Symbol: G0s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008059.3 | NP_032085.1 | 2 | 732 | ||
| GenBank mRNA | AK003165.1 | - | 2 | 654 | ||