siRNA Details
| Gene Symbol: Cyp3a59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105160.1 | NP_001098630.1 | 11 | 1200 | ||
| Gene Symbol: Cyp3a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019792.2 | NP_062766.2 | 11 | 1200 | ||
| GenBank mRNA | AF204959.1 | - | 11 | 1199 | ||
| AK149541.1 | - | 10 | 1113 | |||
| BC028855.1 | - | 11 | 1191 | 5124151 | ||
| Y11995.1 | - | 11 | 1202 | |||

Cyp3a59
Genomic Map