siRNA Details
| Gene Symbol: 9130023H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177001.3 | NP_795975.1 | 1 | 1019 | ||
| GenBank mRNA | AK033604.1 | - | 1 | 979 | ||
| AK084086.1 | - | 1 | 1022 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130023H24Rik| Gene Symbol: 9130023H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177001.3 | NP_795975.1 | 1 | 1019 | ||
| GenBank mRNA | AK033604.1 | - | 1 | 979 | ||
| AK084086.1 | - | 1 | 1022 | |||