siRNA Details
| Gene Symbol: Lce1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028625.2 | NP_082901.1 | 2 | 553 | ||
| GenBank mRNA | AK003412.1 | - | 2 | 554 | ||
| BC062142.1 | - | 2 | 530 | |||
| BC080817.1 | - | 2 | 530 | |||
| BC100540.1 | - | 2 | 540 | |||
| BC147122.1 | - | 2 | 546 | 9056142 | ||

Lce1a2
Genomic Map