siRNA Details
| Gene Symbol: Ankrd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081379.2 | NP_001074848.2 | 9 | 3051 | ||
| GenBank mRNA | AK046451.1 | - | 1 | 154 | ||
| BC158043.1 | - | 8 | 2716 | 9088159 | ||
| BC172141.1 | - | 9 | 2813 | 9054811 | ||

Ankrd11
Genomic Map