siRNA Details
| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134136.3 | NP_598897.2 | 18 | 3129 | ||
| GenBank mRNA | AK031096.1 | - | 18 | 3079 | ||
| AK031347.1 | - | 18 | 3060 | |||
| AK089046.1 | - | 18 | 3098 | |||
| AY267463.1 | - | 18 | 3059 | |||
| BC056348.1 | - | 18 | 3081 | 5721407 | ||

Fbxo38
Genomic Map