siRNA Details
Gene Symbol: Gm15107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081648.1 | NP_001075117.1 | 1825 | |||
GenBank mRNA | BC109131.1 | - | 1794 | 40038981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm15107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081648.1 | NP_001075117.1 | 1825 | |||
GenBank mRNA | BC109131.1 | - | 1794 | 40038981 |