siRNA Details
| Gene Symbol: Nphp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028721.3 | NP_082997.3 | 2 | 706 | ||
| NM_172460.1 | NP_766048.1 | 2 | 706 | |||
| GenBank mRNA | AK048877.1 | - | 2 | 479 | ||
| AK052281.1 | - | 2 | 493 | |||
| AK133217.1 | - | 2 | 707 | |||
| AK173327.1 | - | 2 | 285 | |||
| AY259499.1 | - | 2 | 646 | |||
| BC115724.1 | - | 2 | 236 | 40086844 | ||

Nphp3
Genomic Map