siRNA Details
| Gene Symbol: Cry2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009963.4 | NP_034093.1 | 9 | 1541 | ||
| GenBank mRNA | AB003433.1 | - | 9 | 1426 | ||
| AF156987.1 | - | 9 | 1509 | |||
| AK041696.1 | - | 9 | 1509 | |||
| AK133781.1 | - | 9 | 1516 | |||
| AK172994.1 | - | 9 | 1473 | |||
| BC006077.1 | - | 3 | 449 | |||
| BC054794.1 | - | 9 | 1497 | 6834104 | ||
| BC066799.1 | - | 9 | 1500 | 30103169 | ||

Cry2
Genomic Map