siRNA Details
| Gene Symbol: Hist1h2ag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178186.3 | NP_835493.1 | 1 | 427 | ||
| GenBank mRNA | BC065803.1 | - | 1 | 427 | 1263745 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2ag| Gene Symbol: Hist1h2ag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178186.3 | NP_835493.1 | 1 | 427 | ||
| GenBank mRNA | BC065803.1 | - | 1 | 427 | 1263745 | |