siRNA Details
| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134383.1 | NP_001127855.1 | 3 | 1170 | ||
| NM_001134384.1 | NP_001127856.1 | 3 | 1204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqsec1| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134383.1 | NP_001127855.1 | 3 | 1170 | ||
| NM_001134384.1 | NP_001127856.1 | 3 | 1204 | |||