siRNA Details
| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134383.1 | NP_001127855.1 | 3 | 1690 | ||
| NM_001134384.1 | NP_001127856.1 | 3 | 1724 | |||
| GenBank mRNA | AK220355.1 | - | 1 | 141 | ||
| BC026481.1 | - | 1 | 194 | |||

Iqsec1
Genomic Map