siRNA Details
| Gene Symbol: Zfp655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083958.1 | NP_001077427.1 | 2 | 205 | ||
| NM_028298.3 | NP_082574.1 | 2 | 161 | |||
| GenBank mRNA | AK012336.1 | - | 2 | 164 | ||
| AK018500.1 | - | 2 | 196 | |||
| AK161949.1 | - | 2 | 207 | |||
| AK220552.1 | - | 2 | 166 | |||
| BC019153.1 | - | 2 | 171 | |||
| BC034731.1 | - | 2 | 164 | |||
| BC060660.1 | - | 2 | 197 | 30355779 | ||
| BC061032.1 | - | 2 | 174 | |||
| BC062945.1 | - | 2 | 161 | 30357186 | ||

Zfp655
Genomic Map