siRNA Details
| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111143.1 | NP_001104613.1 | 5 | 591 | ||
| GenBank mRNA | BC147477.1 | - | 5 | 588 | 9056130 | |
| BC171994.1 | - | 5 | 588 | 9054280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cym| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111143.1 | NP_001104613.1 | 5 | 591 | ||
| GenBank mRNA | BC147477.1 | - | 5 | 588 | 9056130 | |
| BC171994.1 | - | 5 | 588 | 9054280 | ||