siRNA Details
| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144880.4 | NP_659129.2 | 10 | 1681 | ||
| GenBank mRNA | AK145397.1 | - | 11 | 1067 | ||
| BC023062.1 | - | 4 | 251 | |||
| BC059026.1 | - | 10 | 1681 | 6810754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r5a| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144880.4 | NP_659129.2 | 10 | 1681 | ||
| GenBank mRNA | AK145397.1 | - | 11 | 1067 | ||
| BC023062.1 | - | 4 | 251 | |||
| BC059026.1 | - | 10 | 1681 | 6810754 | ||