siRNA Details
| Gene Symbol: 5031414D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198642.2 | NP_941044.3 | 4 | 978 | ||
| GenBank mRNA | AK170426.1 | - | 4 | 942 | ||
| BC052668.1 | - | 4 | 990 | 30054870 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5031414D18Rik| Gene Symbol: 5031414D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198642.2 | NP_941044.3 | 4 | 978 | ||
| GenBank mRNA | AK170426.1 | - | 4 | 942 | ||
| BC052668.1 | - | 4 | 990 | 30054870 | ||